missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-19093
This item is not returnable.
View return policy
Description
SF2 Polyclonal specifically detects SF2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| SF2 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| Alternative-splicing factor 1, ASFpre-mRNA-splicing factor SF2, P33 subunit, MGC5228, serine/arginine-rich splicing factor 1, SF2FLJ53078, SF2p33, SFRS1splicing factor 2, Splicing factor, arginine/serine-rich 1ASF-1, SR splicing factor 1, SRp30a | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| NP_008855.1 | |
| SRSF1 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human alternative splicing factor (splicing factor, arginine/serine-rich 1) using the numbering given in entry NP_008855.1 (GeneID 6426). | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 6426 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction