missing translation for 'onlineSavingsMsg'
Learn More

SF2 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18400555 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400555 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400555 Supplier Novus Biologicals Supplier No. NBP119093IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SF2 Polyclonal antibody specifically detects SF2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SF2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Alternative-splicing factor 1, ASFpre-mRNA-splicing factor SF2, P33 subunit, MGC5228, serine/arginine-rich splicing factor 1, SF2FLJ53078, SF2p33, SFRS1splicing factor 2, Splicing factor, arginine/serine-rich 1ASF-1, SR splicing factor 1, SRp30a
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human alternative splicing factor (splicing factor, arginine/serine-rich 1) using the numbering given in entry NP_008855.1 (GeneID 6426).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6426
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.