missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SF 11

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SF 11 is an NPY Y2 antagonist (IC50 = 199 nM). Displays no affinity for Y1 receptor at concentrations up to 35 μM. Causes significant lethality in mice. Displays antidepressant effects in rats. Brain penetrant.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 446.6 |
| Purity | >99% |
| Molecular Weight (g/mol) | 446.609 |
| Chemical Name or Material | SF 11 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction