Learn More
Vapourtec Ltd. SF-10 Tubing Kit for Solvent/Reagent Valve
Product Code. 15988082
Encompass
Click to view available options
For Use With:
SF-10 Reagent Pump
Product Code. 15988082
Brand: Vapourtec Ltd. 501512
This item requires a price update prior to being ordered.
Request a Price for this item here.
The innovative SF-10 is a highly chemically resistant pump capable of generating a smooth, continuous flow. The flow rate does not fluctuation like a conventional peristaltic pump offering greater precision. The SF-10 is highly versatile, offering five operating modes (constant flow rate, ramping flow rate, metering gases, volume dispensing and precision pressure regulator) in the one compact unit
  • Tubing kit to connect solvent/reagent valve to the SF-10 reagent pump
Specifications
For Use With SF-10 Reagent Pump