missing translation for 'onlineSavingsMsg'
Learn More

SF-1/NR5A1/Steroidogenic Factor 1 Antibody (NR5A1/4368R), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18326754 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326754 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326754 Supplier Novus Biologicals Supplier No. NBP314291JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SF-1/NR5A1/Steroidogenic Factor 1 Monoclonal antibody specifically detects SF-1/NR5A1/Steroidogenic Factor 1 in Human, Rat samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SF-1/NR5A1/Steroidogenic Factor 1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NR5A1/4368R
Conjugate Janelia Fluor 646
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AD4BPSTF-1, Adrenal 4-binding protein, ELP, FTZ1adrenal 4 binding protein, FTZF1nuclear receptor AdBP4, Fushi tarazu factor homolog 1, Nuclear receptor subfamily 5 group A member 1, nuclear receptor subfamily 5, group A, member 1, SF-1POF7, SF1steroidogenic factor 1, Steroid hormone receptor Ad4BP, steroidogenic factor-1
Host Species Rabbit
Immunogen Recombinant fragment of human SF-1/NR5A1/Steroidogenic Factor 1 protein (exact sequence is proprietary) (Uniprot: Q13285)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 2516
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.