missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SEZ6/BSRP-C Antibody (Abbv-011) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | SEZ6/BSRP-C |
|---|---|
| Clone | Abbv-011 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30048190
|
Novus Biologicals™
NBP3-28127-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046005
|
Novus Biologicals™
NBP3-28127-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044418
|
Novus Biologicals™
NBP3-28127-100ug |
100 μg |
£266.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SEZ6/BSRP-C Monoclonal antibody specifically detects SEZ6/BSRP-C in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| SEZ6/BSRP-C | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Human | |
| hSEZ-6, seizure related 6 homolog (mouse) | |
| SEZ6 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| Abbv-011 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q53EL9 | |
| 124925 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title