missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Sewage Effluent Kit

Product Code. 11918976
Change view
Click to view available options
Includes:
3 x 100mL Sample Containers, 2 x 10mL Test Tubes, 1 x 13′′ Turbidity Tube, 1 x Thermometer, 1 x Diskette Comparator, 1 x Test Tube Brush, Reagents for Acidifying SE, Permanganate Value and Universal pH
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
11918976 3 x 100mL Sample Containers, 2 x 10mL Test Tubes, 1 x 13′′ Turbidity Tube, 1 x Thermometer, 1 x Diskette Comparator, 1 x Test Tube Brush, Reagents for Acidifying SE, Permanganate Value and Universal pH Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11918976 Supplier Palintest™ Supplier No. SP304
  • ONLINE EXCLUSIVE PRICE
    £378.63
    £323.30 / Pack of 1
    Save £55.33
    14% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

For Use With (Application) Drinking water, wastewater and process water product range
Includes 3 x 100mL Sample Containers, 2 x 10mL Test Tubes, 1 x 13′′ Turbidity Tube, 1 x Thermometer, 1 x Diskette Comparator, 1 x Test Tube Brush, Reagents for Acidifying SE, Permanganate Value and Universal pH
Type Sewage effluent kit
compliance-icons compliance-icons compliance-icons
Product Identifier
  • photometry test kit sulphate 0-200PPM
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash contaminated skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P391-Collect spillage.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see medical advice on this label).
  • P363-Wash contaminated clothing before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: potassium permanganate, potassium hydrogen sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.