missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ SEW 2871
Description
Novel, potent and selective sphingosine-1-phosphate 1 (S1P1) receptor agonist. Activates S1P1 receptor with an EC50 of 13 nM, but does not activate S1P2, S1P3, S1P4 or S1P5 receptors at concentrations up to 10 μM. Cell-permeable and active in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Product Line | Tocris |
| Formula Weight | 440.36 |
| Purity | >99% |
| Molecular Weight (g/mol) | 440.36 |
| Chemical Name or Material | SEW 2871 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?