missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETMAR Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25814-1-AP-20UL
This item is not returnable.
View return policy
Description
Potential coactivator of the androgen receptor. Mediates STAT3 degradation. May play critical roles in two RAB6-dependent retrograde transport processes: one from endosomes to the Golgi and the other from the Golgi to the ER. This protein binds the HIV-1 TATA element and inhibits transcriptional activation by the TATA-binding protein (TBP).Specifications
| SETMAR | |
| Polyclonal | |
| Unconjugated | |
| SETMAR | |
| HsMar1, METNASE, SETMAR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6419 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q53H47 | |
| SETMAR | |
| SETMAR Fusion Protein Ag22938 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction