missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SETMAR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 720343
This item is not returnable.
View return policy
Description
This antibody is predicted to react with Monkey, Bovine, Sheep.
SETMAR (SET domain and mariner transposase fusion gene) is a histone methyltransferase that methylates Lys-4 and Lys-36 of histone H3, creating specific tags for epigenetic transcriptional activation. It specifically mediates dimethylation of H3 Lys-36. SETMAR has sequence-specific DNA-binding activity, and recognizes the 19-mer core of the 5'-terminal inverted repeats (TIRs) of the Hsmar1 transposable element. SETMAT has DNA nicking activity, and in vivo end-joining activity, and may mediate genomic integration of foreign DNA.
Specifications
| SETMAR | |
| Polyclonal | |
| Unconjugated | |
| SETMAR | |
| 5830404F24Rik; epidermal tumor expressed transcript 2; Etet2; Histone-lysine N-methyltransferase; Histone-lysine N-methyltransferase SETMAR; Mar1; METNASE; SET domain and mariner transposase fusion gene; SET domain and mariner transposase fusion gene-containing protein; SET domain and mariner transposase fusion gene-containing protein homolog; SET domain and mariner transposase fusion protein; SET domain and mariner transposase fusion protein homolog; SET domain without mariner transposase fusion; SET domain without mariner transposase fusion protein; Setmar; Transposon Hsmar1 transposase | |
| Rabbit | |
| Protein G | |
| RUO | |
| 6419, 74729 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4 | |
| Q53H47, Q80UJ9 | |
| SETMAR | |
| Protein corresponding to human SETMAR [aa15-aa303]. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction