missing translation for 'onlineSavingsMsg'
Learn More

Setiptiline maleate, MedChemExpress

Product Code. 16453957
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453957

Brand: Medchem Express HY32329A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Setiptiline maleate (MO-8282 maleate) is a serotonin receptor antagonist.

Setiptiline maleate is a tetracyclic antidepressant (TeCA) which acts as a noradrenergic and specific serotonergic antidepressant (NaSSA). Setiptiline maleate acts as a norepinephrine reuptake inhibitor, α2-adrenergic receptor antagonist, and serotonin receptor antagonist, likely at the 5-HT2A, 5-HT2C, and/or 5-HT3 subtypes, as well as an H1 receptor inverse agonist/antihistamine.

Specifications

Chemical Name or Material Setiptiline maleate
CAS 85650-57-3
Color White
Purity Grade Notes Research
Molecular Formula C23H23NO4
Quantity 50 mg
Synonym MO-8282
Solubility Information DMSO : 100 mg/mL (264.95 mM; Need ultrasonic)
SMILES CN1CC(C(C=CC=C2)=C2C3)=C(CC1)C4=C3C=CC=C4.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 377.43
Formula Weight 377.43
Percent Purity 98.18%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.