missing translation for 'onlineSavingsMsg'
Learn More

Setipiprant, MedChemExpress

Product Code. 16447098
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16447098

Brand: Medchem Express HY16635

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Setipiprant is an orally available, selective CRTH2 antagonist.

Setipiprant is an orally available, selective CRTH2 antagonist.

Specifications

Chemical Name or Material Setipiprant
CAS 866460-33-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H19FN2O3
Quantity 10 mg
Synonym ACT-129968 KYTH-105
Solubility Information DMSO : ≥ 36 mg/mL (89.46 mM)
SMILES O=C(O)CN1C2=C(CN(C(C3=C4C=CC=CC4=CC=C3)=O)CC2)C5=C1C=CC(F)=C5
Molecular Weight (g/mol) 402.42
Formula Weight 402.42
Percent Purity 98.7%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.