missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETDB2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38422-100ul
This item is not returnable.
View return policy
Description
SETDB2 Polyclonal antibody specifically detects SETDB2 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| SETDB2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| C13orf4, chromosome 13 open reading frame 4, chronic lymphocytic leukemia deletion region 8, Chronic lymphocytic leukemia deletion region gene 8 protein, CLLD8DKFZp761J1217, DKFZp586I0123, EC 2.1.1.43, histone-lysine N-methyltransferase SETDB2, KMT1FCLLL8, Lysine N-methyltransferase 1F, SET domain bifurcated 2, SET domain, bifurcated 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 148-378 of human SETDB2 (NP_114121.2).,, Sequence:, SGACLMKMPLNLKGENPLQLPIKCHFQRRHAKTNSHSSALHVSYKTPCGRSLRNVEEVFRYLLETECNFLFTDNFSFNTYVQLARNYPKQKEVVSDVDISNGVESVPISFCNEIDSRKLPQFKYRKTVWPRAYNLTNFSSMFTDSCDCSEGCIDITKCACLQLTARNAKTSPLSSDKITTGYKYKRLQRQIPTGIYECSLLCKCNRQLCQNRVVQHGPQVRLQVFKTEQKG | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 83852 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction