missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETDB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | SETDB1 |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16898163
|
Proteintech
11231-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16888163
|
Proteintech
11231-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human SETDB1.The calculated molecular weight of SETDB1 is 143 kDa, but the modified SETDB1 protein is about 170 kDa.
This gene encodes a histone methyltransferase. The encoded enzyme catalyzes the reaction of S-adenosyl-L-methionine and histone L-lysine to produce S-adenosyl-L-homocysteine and histone N (6)-methyl-L-lysine. The encoded protein likely functions in transcriptional repression. Alternatively spliced transcript variants have been described.Specifications
| SETDB1 | |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| O88974, Q15047 | |
| 689883, 84505, 9869 | |
| SETDB1 Fusion Protein Ag1725 | |
| Primary | |
| -20°C | |
| SETDB1 |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ESET, H3 K9 HMTase 4, H3 K9 HMTase4, KG1T, KIAA0067, KMT1E, Lysine N methyltransferase 1E, SET domain bifurcated 1, SET domain, bifurcated 1, SETDB1 | |
| Setdb1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title