missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SETDB1 Monoclonal Antibody (K.137.7)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA514960
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other SET-domain containing histone methyltransferase proteins.
This gene encodes a histone methyltransferase. The encoded enzyme catalyzes the reaction of S-adenosyl-L-methionine and histone L-lysine to produce S-adenosyl-L-homocysteine and histone N(6)-methyl-L-lysine. The encoded protein likely functions in transcriptional repression. Alternatively spliced transcript variants have been described.
Specifications
| SETDB1 | |
| Monoclonal | |
| 9 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| Q15047 | |
| SETDB1 | |
| Synthetic peptide corresponding to the carboxy terminus of the human ESET protein. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
| ChIP Assay, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| K.137.7 | |
| Unconjugated | |
| SETDB1 | |
| AU022152; ERG-associated protein with a SET domain, ESET; ERG-associated protein with SET domain; ESET; H3-K9-HMTase 4; H3-K9-HMTase4; histone H3-K9 methyltransferase 4; Histone-lysine N-methyltransferase SETDB1; histone-lysine N-methyltransferase, H3lysine-9 specific 4; KG1T; KIAA0067; KMT1E; Lysine N-methyltransferase 1E; mKIAA0067; SET domain bifurcated 1; SET domain ERG-associated histone methyltransferase; SET domain, bifurcated 1; SETDB1; TDRD21; tudor domain containing 21 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9869 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction