missing translation for 'onlineSavingsMsg'
Learn More

SETD8 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18325732 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325732 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325732 Supplier Novus Biologicals Supplier No. NBP299863IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SETD8 Polyclonal specifically detects SETD8 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD8
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias H4-K20-HMTase SETD8, H4K20-specific histone methyltransferase splice variant Set8b, Histone-lysine N-methyltransferase SETD8, KMT5A, KMT5A SET domain-containing protein 8, Lysine N-methyltransferase 5A, PR/SET domain containing protein 8, PR/SET domain-containing protein 07, PR/SET07, PRSET7, PR-Set7EC 2.1.1.43, SET domain containing (lysine methyltransferase) 8, SET07SET8H4-K20-specific histone methyltransferase, SET8
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant SETD8 (NP_065115.3; Lys195-His352).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 387893
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.