missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SETD8 Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 387893 |
| Concentration | 1 mg/ml |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP1-91269. For further blocking peptide related protocol, click here . |
| Gene Alias | H4-K20-HMTase SETD8, H4K20-specific histone methyltransferase splice variant Set8b, Histone-lysine N-methyltransferase SETD8, KMT5A, KMT5A SET domain-containing protein 8, Lysine N-methyltransferase 5A, PR/SET domain containing protein 8, PR/SET domain-containing protein 07, PR/SET07, PRSET7, PR-Set7EC 2.1.1.43, SET domain containing (lysine methyltransferase) 8, SET07SET8H4-K20-specific histone methyltransferase, SET8 |
| Gene Symbol | KMT5A |
| Quantity | 0.1 mg |
| Research Category | Cell Cycle and Replication, Epigenetics |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?