missing translation for 'onlineSavingsMsg'
Learn More

SETD7 Antibody (RAB-C220) [Janelia Fluor™ 585], Novus Biologicals™

Product Code. 30163880
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30163880 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30163880 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP320145JF585

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SETD7 Monoclonal antibody specifically detects SETD7 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Western Blot,Mass Spectrometry,ChIP assay (ChIP),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD7
Applications Flow Cytometry, ELISA, Immunoprecipitation, Western Blot, Mass Spectrometry, ChIP Assay, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone RAB-C220
Conjugate Janelia Fluor 585
Formulation 50mM Sodium Borate
Gene Alias EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9
Host Species Rabbit
Immunogen This antibody was obtained by recombinant antibody (rAb) phage display recognizing SETD7 protein under non-denaturing conditions.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 80854
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.