missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD7 Antibody (RAB-C220), Alexa Fluor™ 405 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20145AF405
This item is not returnable.
View return policy
Description
SETD7 Monoclonal antibody specifically detects SETD7 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
| SETD7 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing SETD7 protein under non-denaturing conditions. | |
| 0.1 mL | |
| Epigenetics | |
| 80854 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Flow Cytometry, ELISA, Immunofluorescence | |
| RAB-C220 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction