missing translation for 'onlineSavingsMsg'
Learn More

SETD7/9 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18370266 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μg
50 μg
Unit Size:
25µL
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370266 50 μg 50µg
18346983 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18370266 Supplier Novus Biologicals Supplier No. NBP31867550UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SETD7/9 Polyclonal antibody specifically detects SETD7/9 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD7/9
Applications Western Blot
Classification Polyclonal
Concentration 2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS
Gene Accession No. Q8WTS6
Gene Alias EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human SETD7/9 (SET domain containing (lysine methyltransferase) 9), using a recombinant protein.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Histones and Modified Histones, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 80854
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.