missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SETD7/9 Polyclonal antibody specifically detects SETD7/9 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | SETD7/9 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 |
| Formulation | PBS |
| Gene Accession No. | Q8WTS6 |
| Gene Alias | EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?