missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD7/9 Antibody (s4E5), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04298PEATT594
This item is not returnable.
View return policy
Description
SETD7/9 Monoclonal specifically detects SETD7/9 in Human samples. It is validated for Flow Cytometry.
Specifications
| SETD7/9 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| SETD7 | |
| Recombinant human SET7/9 (1-366aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Anti-human SET7/9 mAb, clone s4E5, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with recombinant human SET7/9. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| s4E5 | |
| Flow Cytometry | |
| EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9 | |
| Mouse | |
| Protein G purified | |
| Histones and Modified Histones, Signal Transduction | |
| 80854.0 | |
| Human | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction