missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD7/9 Antibody (19), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06624-50ul
This item is not returnable.
View return policy
Description
SETD7/9 Monoclonal antibody specifically detects SETD7/9 in Human samples. It is validated for Western Blot
Specifications
| SETD7/9 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 19 | |
| Western Blot 1:500-1:1000 | |
| EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SETD7/9 (Accession#: NP_085151.1; Asp2-Lys366). | |
| 50 μg | |
| Histones and Modified Histones, Signal Transduction | |
| 80854 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction