missing translation for 'onlineSavingsMsg'
Learn More

SETD7/9 Antibody (001), DyLight 488 , Novus Biologicals™

Product Code. 18422829 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422829 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422829 Supplier Novus Biologicals Supplier No. NBP289842G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SETD7/9 Monoclonal antibody specifically detects SETD7/9 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD7/9
Applications ELISA
Classification Monoclonal
Clone 001
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human SETD7/9 (NP_085151.1; Asp2-Lys366).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Histones and Modified Histones, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 80854
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.