missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SETD6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577180
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
SETD6 (SET domain-containing protein 6) is a 473 amino acid protein that belongs to the SETD6 family. Containing one SET domain, SETD6 is structurally similar to the Rubisco large subunit methyltransferase. SETD6 is a proteinlysine N-methyltransferase that specifically monomethylates Lys-310 of the NFκB p65 subunit of NFκB complex, leading to down-regulate NFκB transcription factor activity. SETD6-mediated methylation renders NFκB p65 inert and attenuates NFκB p65-driven transcriptional programs, including inflammatory responses in primary immune cells. The SETD6-initiated lysine-methylation signaling cascade acts to restrain activation of NFκB-mediated inflammatory responses in diverse cell types.
Spécification
| SETD6 | |
| Polyclonal | |
| Unconjugated | |
| SETD6 | |
| 0610039J04Rik; 3110004G14Rik; AI413388; C76402; N-lysine methyltransferase SETD6; RGD1560538; SET domain containing 6; SET domain-containing protein 6; SETD6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 291844, 66083, 79918 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| D3ZSK5, Q8TBK2, Q9CWY3 | |
| SETD6 | |
| Recombinant full length Human SETD6. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu