missing translation for 'onlineSavingsMsg'
Learn More

SETD3 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18414547 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414547 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414547 Supplier Novus Biologicals Supplier No. NBP232137JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SETD3 Polyclonal antibody specifically detects SETD3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD3
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias C14orf154, chromosome 14 open reading frame 154, DKFZp761E1415, EC 2.1.1.43, FLJ23027, MGC87236, SET domain containing 3, SET domain-containing protein 3
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 544 to 594 of human SET Domain Containing 3 using the numbering given in entry NP_115609.2 (GeneID 84193).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84193
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.