missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD3 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32136UV
This item is not returnable.
View return policy
Description
SETD3 Polyclonal antibody specifically detects SETD3 in Human, Mouse samples. It is validated for Western Blot
Specifications
| SETD3 | |
| Polyclonal | |
| Western Blot | |
| C14orf154, chromosome 14 open reading frame 154, DKFZp761E1415, EC 2.1.1.43, FLJ23027, MGC87236, SET domain containing 3, SET domain-containing protein 3 | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human SET Domain Containing 3 using the numbering given in entry NP_115609.2 (GeneID 84193). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 84193 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction