missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SETD1B Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA540072

Product Code. 15556761

  • £443.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SETD1B Polyclonal Antibody for Western Blot, ELISA

Two distinct signaling pathways activate the host innate immunity against viral infection. One pathway is reliant on members of the Toll-like receptor (TLR) family while the other uses the RNA helicase RIG-I as a receptor for intracellular viral double-stranded RNA as a trigger for the immune response. VISA is a mitochondrial membrane protein that was identified as a critical component in the IFN-b signaling pathways that recruits IRF-3 to RIG-I, leading to its activation and that of NF-kappa-B. VISA is also thought to interact with other components of the innate immune pathway such as the TLR adapter protein TRIF, TRAF2 and TRAF6. VISA also interacts with the IKK-a, IKK-b and IKK-iota kinases through its C-terminal region. Cleavage of this region by the Hepatitis C virus (HCV) protease allows HCV to escape the host immune system. At least three isoforms of VISA are known to exist.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SETD1B
Polyclonal
Unconjugated
Setd1b
AA516740; BC035291; Histone-lysine N-methyltransferase SETD1B; hSET1B; Kiaa1076; KMT2G; LOC100359816; Lysine N-methyltransferase 2G; mKIAA1076; rCG21620-like; SET domain containing 1B; SET domain-containing protein 1B; SET1B; SETD1B
Rabbit
100 μL
Primary
Mouse
Antibody
IgG
ELISA, Western Blot
Conc. Not Determined
Whole serum with 0.05% sodium azide
Q8CFT2
Setd1b
KLH-conjugated synthetic peptides containing sequences from the central part of the protein corresponding to mouse Setd1b (Set domain containing 1B).
RUO
208043
-20°C or -80°C if preferred
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.