missing translation for 'onlineSavingsMsg'
Learn More

SETD1A Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18357856 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357856 100 μg 100µL
18315906 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18357856 Supplier Novus Biologicals Supplier No. NBP318681100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SETD1A Polyclonal antibody specifically detects SETD1A in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETD1A
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100
Formulation Whole antiserum
Gene Accession No. O15047
Gene Alias histone-lysine N-methyltransferase SETD1A, hSET1A, KIAA0339EC 2.1.1.43, KMT2FSET1, Lysine N-methyltransferase 2F, SET domain containing 1A, SET domain-containing protein 1A, Set1, Set1/Ash2 histone methyltransferase complex subunit SET1, SET1A
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against mouse SETD1A (Set domain containing 1A), using three KLH-conjugated synthetic peptides: two containing an amino acid sequence from the N-terminal part of the protein and one containing an amino acid sequence from the central part of the protein (1).
Purification Method Whole Antiserum
Quantity 100 μg
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 9739
Target Species Human, Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.