missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SETD1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563563
This item is not returnable.
View return policy
Description
Immunogen sequence: SQFRSSDANY PAYYESWNRY QRHTSYPPRR ATREEPPGAP FAENTAERFP PSYTSYLPPE PSRPTDQDYR PPASEA Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
SET1A is a component of a histone methyltransferase (HMT) complex that produces mono-, di-, and trimethylated histone H3 at Lys4. The complex is the analog of the S. cerevisiae Set1/COMPASS complex.
Specifications
| SETD1A | |
| Polyclonal | |
| Unconjugated | |
| SETD1A | |
| BC010250; Histone-lysine N-methyltransferase SETD1A; hSET1A; KIAA0339; KMT2F; Lysine N-methyltransferase 2F; mKIAA0339; mNSC1; non-selective cation channel 1; Nsccn1; SET domain containing 1A; SET domain-containing protein 1A; SET1; Set1/Ash2 histone methyltransferase complex subunit SET1; SET1A; SETD1A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9739 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15047 | |
| SETD1A | |
| Recombinant protein corresponding to Human SETD1A. Recombinant protein control fragment (Product #RP-104165). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction