missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35807-20ul
This item is not returnable.
View return policy
Description
SETD1A Polyclonal antibody specifically detects SETD1A in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| SETD1A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| histone-lysine N-methyltransferase SETD1A, hSET1A, KIAA0339EC 2.1.1.43, KMT2FSET1, Lysine N-methyltransferase 2F, SET domain containing 1A, SET domain-containing protein 1A, Set1, Set1/Ash2 histone methyltransferase complex subunit SET1, SET1A | |
| Recombinant protein of human SETD1A. | |
| 20 μL | |
| DNA Repair | |
| 9739 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction