missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SET1B Polyclonal antibody specifically detects SET1B in Human samples. It is validated for Immunofluorescence
Specifications
Specifications
| Antigen | SET1B |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.1.1, FLJ20803, histone-lysine N-methyltransferase SETD1B, hSET1B, KIAA1076EC 2.1.1.43, KMT2GSET1B, Lysine N-methyltransferase 2G, SET domain containing 1B, SET domain-containing protein 1B, Set1B |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human SET1B fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?