missing translation for 'onlineSavingsMsg'
Learn More

SET1B Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18341872 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341872 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341872 Supplier Novus Biologicals Supplier No. NBP306532AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SET1B Polyclonal antibody specifically detects SET1B in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SET1B
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias EC 2.1.1, FLJ20803, histone-lysine N-methyltransferase SETD1B, hSET1B, KIAA1076EC 2.1.1.43, KMT2GSET1B, Lysine N-methyltransferase 2G, SET domain containing 1B, SET domain-containing protein 1B, Set1B
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SET1B fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23067
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.