missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SET1B Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28685AF532
This item is not returnable.
View return policy
Description
SET1B Polyclonal antibody specifically detects SET1B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SET1B | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 2.1.1, FLJ20803, histone-lysine N-methyltransferase SETD1B, hSET1B, KIAA1076EC 2.1.1.43, KMT2GSET1B, Lysine N-methyltransferase 2G, SET domain containing 1B, SET domain-containing protein 1B, Set1B | |
| The immunogen recognized by this antibody maps to a region between residue 1400 and 1450 of human SET domain containing 1B using the numbering given in entry NP_055863.1 (GeneID 23067). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23067 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction