missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SET Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55201-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to SET. It can recognize isoform1 and isoform2 of SET.
SET is a multitasking protein, involved in apoptosis, transcription, nucleosome assembly and histone binding. Isoform 2 anti-apoptotic activity is mediated by inhibition of the GZMA-activated DNase, NME1.Specifications
| SET | |
| Polyclonal | |
| Unconjugated | |
| SET | |
| 2PP2A, HLA DR associated protein II, I 2PP2A, I2PP2A, IGAAD, IPP2A2, PHAPII, Protein SET, SET, SET nuclear oncogene, TAF I, TAF IBETA, Template activating factor I | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 307947, 56086, 6418 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q01105, Q63945, Q9EQU5 | |
| SET | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction