missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SET Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544576
This item is not returnable.
View return policy
Description
Peptide sequence: IDEVQNEIDR LNEQASEEIL KVEQKYNKLR QPFFQKRSEL IAKIPNFWVT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
SET is a multitasking protein, involved in apoptosis, transcription, nucleosome assembly and histone binding. Isoform 2 anti-apoptotic activity is mediated by inhibition of the GZMA-activated DNase, NME1.
Specifications
| SET | |
| Polyclonal | |
| Unconjugated | |
| SET | |
| 2610030F17Rik; 2PP2A; 5730420M11Rik; AA407739; Ab1-115; HLA-DR-associated protein II; hypothetical protein LOC538444; I IGAAD; I2PP2A; I-2PP2A; IGAAD; inhibitor of granzyme A-activated DNase; inhibitor-2 of protein phosphatase-2A; IPP2A2; Liver regeneration-related protein LRRGR00002; OTTHUMP00000022286; PHAPII; Phosphatase 2A inhibitor I2PP2A; protein phosphatase 2A inhibitor 2; protein phosphatase type 2A inhibitor; protein SET; RP11-216B9.3; Set; SET nuclear oncogene; SET nuclear proto-oncogene; SET translocation; SET translocation (myeloid leukemia-associated); SET-like protein; StF-IT-1; TAF-1a; TAF-1b; TAF-I; TAF-IBETA; Template-activating factor I; Template-Activating Factor-I, chromatin remodelling factor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56086, 6418 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q01105, Q9EQU5 | |
| SET | |
| Synthetic peptide directed towards the N-terminal of human SET (aa 48-97). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction