missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SET Mouse anti-Human, Clone: PCRP-SET-1C6, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07837-20UG
This item is not returnable.
View return policy
Description
SET Monoclonal specifically detects SET in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
Specifications
| SET | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
| 2PP2A, HLA-DR-associated protein II, I2PP2A, I-2PP2A, IGAAD, Inhibitor of granzyme A-activated DNase, inhibitor-2 of protein phosphatase-2A, IPP2A2, PHAPIITAF-I, Phosphatase 2A inhibitor I2PP2A, protein phosphatase type 2A inhibitor, protein SET, SET nuclear oncogene, SET translocation (myeloid leukemia-associated), TAF-IBETA, Template-activating factor I, Template-Activating Factor-I, chromatin remodelling factor | |
| Recombinant full-length human SET protein | |
| 20 μg | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 6418 | |
| Store at 4°C. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array | |
| PCRP-SET-1C6 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction