missing translation for 'onlineSavingsMsg'
Learn More

SET Antibody (PCRP-SET-1C6) - Azide and BSA Free, Novus Biologicals™

Product Code. 18386383 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386383 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386383 Supplier Novus Biologicals Supplier No. NBP308730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SET Monoclonal specifically detects SET in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SET
Applications Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array
Classification Monoclonal
Clone PCRP-SET-1C6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1-2 ug/mL, Flow Cytometry : 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence : 1-2 ug/mL, Protein Array
Formulation 10mM PBS
Gene Alias 2PP2A, HLA-DR-associated protein II, I2PP2A, I-2PP2A, IGAAD, Inhibitor of granzyme A-activated DNase, inhibitor-2 of protein phosphatase-2A, IPP2A2, PHAPIITAF-I, Phosphatase 2A inhibitor I2PP2A, protein phosphatase type 2A inhibitor, protein SET, SET nuclear oncogene, SET translocation (myeloid leukemia-associated), TAF-IBETA, Template-activating factor I, Template-Activating Factor-I, chromatin remodelling factor
Host Species Mouse
Immunogen Recombinant full-length human SET protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6418
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.