missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SET Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals™ NBP3-30918-100ug

 View more versions of this product

Product Code. 30067212

  • £386.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SET Polyclonal antibody specifically detects SET in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SET
Polyclonal
Western Blot : 1:400
2PP2A, HLA-DR-associated protein II, I2PP2A, I-2PP2A, IGAAD, Inhibitor of granzyme A-activated DNase, inhibitor-2 of protein phosphatase-2A, IPP2A2, PHAPIITAF-I, Phosphatase 2A inhibitor I2PP2A, protein phosphatase type 2A inhibitor, protein SET, SET nuclear oncogene, SET translocation (myeloid leukemia-associated), TAF-IBETA, Template-activating factor I, Template-Activating Factor-I, chromatin remodelling factor
Synthetic peptide derived from the human SET protein
100 μg
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
6418
Store at -70°C. Avoid freeze/thaw cycles.
IgG
Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Antigen Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.