missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Set of Adjustable Feet for UN75plus
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN75plus Oven
Unit Size:
Each
Description
- Includes 4 pieces
- Accessory
Specifications
Specifications
| For Use With (Equipment) | UN75plus Oven |
| Type | Adjustable Feet Set |
| Includes | 4 Pieces |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction