missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Set of 4 dNTPs (100mm)

Product Code. p-7083057 Shop All MP Biomedicals Products
Click to view available options
Quantity:
4 x 100 μmol
4 x 25 μmol
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 11457110

Brand: MP Biomedicals™ 11NTACG100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Offers maximum flexibility in preparation of reaction mixes. MP Biomedicals Set of 4 dNTPs (100mm) includes four separate vials of highly purified sodium salts of dATP, dCTP, dGTP and dTTP. These solutions achieve sensitive and consistent PCR results.

This product allows the addition of each dNTP to be altered as required for specific experiments, such as:

  • When adding a radioactive nucleotide for labeling by PCR
  • To increase the chance of non-specific mutations
  • In PCR, real-time PCR, cDNA synthesis and DNA sequencing
TRUSTED_SUSTAINABILITY

Specifications

Concentration 100 mM
Content And Storage Set 4 DNTPs composed of four separate vials of sodium salts of dATP; dCTP; dGTP and dTTP. Store at -20°C.
Purity or Quality Grade 99+%
Quantity 4 x 25 μmol
For Use With (Application) Standard PCR, Real Time PCR (qPCR), RT-PCR
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.