missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sestrin 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21346-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the sestrin family of PA26-related proteins. The encoded protein may function in the regulation of cell growth and survival. This protein may be involved in cellular response to different stress conditions.Specifications
| Sestrin 2 | |
| Polyclonal | |
| Unconjugated | |
| SESN2 | |
| HI95, SES2, SESN2, SEST2, Sestrin 2, Sestrin2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 230784, 83667 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P58004, P58043 | |
| SESN2 | |
| Sestrin 2 Fusion Protein Ag15669 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction