missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SESN2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £429.00
Specifications
| Antigen | SESN2 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18676858
|
Novus Biologicals
NBP2-41302-0.025mg |
0.025 mg |
£150.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18181939
|
Novus Biologicals
NBP2-41302 |
0.1 mg |
£429.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SESN2 Polyclonal specifically detects SESN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SESN2 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp761M0212, DKFZp761M02121, Hi95, HI95sestrin-2, SES2, SEST2hypoxia induced gene 95, sestrin 2 | |
| SESN2 | |
| IgG | |
| Affinity Purified | |
| 53 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS with 0.02% Sodium Azide | |
| 83667 | |
| Antibody was raised against a 19 amino acid peptide near the center of human SESTRIN2. The immunogen is located within amino acids 220 - 270 of SESTRIN2. Amino Acid Squence: APTPPSEQSSPPSRDPLNN | |
| Primary | |
| At least two isoforms of SESTRIN2 are known to exist; this antibody will detect both isoforms. SESTRIN2 antibody is predicted to not cross-react with SESTRIN1. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title