missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SESN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111712
This item is not returnable.
View return policy
Description
Immunogen sequence: ASRFEIEKRE SMFVFSSDDE EVTPARAVSR HFEDTSYGYK DFSRHGMHVP TFRVQDYC Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 95%.
This gene encodes a member of the sestrin family. Sestrins are induced by the p53 tumor suppressor protein and play a role in the cellular response to DNA damage and oxidative stress. The encoded protein mediates p53 inhibition of cell growth by activating AMP-activated protein kinase, which results in the inhibition of the mammalian target of rapamycin protein. The encoded protein also plays a critical role in antioxidant defense by regenerating overoxidized peroxiredoxins, and the expression of this gene is a potential marker for exposure to radiation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| SESN1 | |
| Polyclonal | |
| Unconjugated | |
| Sesn1 | |
| 1110002G11Rik; AU044290; LOW QUALITY PROTEIN: sestrin-1; p53 activated gene 26; p53 regulated PA26 nuclear protein; p53-regulated protein PA26; PA26; Sesn1; Sest1; sestrin 1; sestrin-1; si:ch211-51a19.4; WITHDRAWN: zgc:103669; zgc:103669; zgc:91970 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27244 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y6P5 | |
| Sesn1 | |
| Recombinant Protein corresonding to Human SESN1. Recombinant protein control fragment (Product #RP-107941). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction