missing translation for 'onlineSavingsMsg'
Learn More

Sesamin, TRC

Product Code. 30410018
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
Product Code. Quantity unitSize
30410018 1 mg 1mg
30428165 10 mg 10mg
30423592 25 mg -
3 options
This item is not returnable. View return policy

Product Code. 30410018

Brand: Toronto Research Chemicals TRCS2805001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-80-7
Formula Weight 354.1103
InChI Formula InChI=1S/C20H18O6/c1-3-15-17(25-9-23-15)5-11(1)19-13-7-22-20(14(13)8-21-19)12-2-4-16-18(6-12)26-10-24-16/h1-6,13-14,19-20H,7-10H2/t13-,14-,19+,20+/m0/s1
IUPAC Name 5-[(3S,3aR,6S,6aR)-6-(1,3-benzodioxol-5-yl)-1,3,3a,4,6,6a-hexahydrofuro[3,4-c]furan-3-yl]-1,3-benzodioxole
Molecular Formula C20 H18 O6
Molecular Weight (g/mol) 354.35
SMILES C1Oc2ccc(cc2O1)[C@H]3OC[C@H]4[C@@H]3CO[C@@H]4c5ccc6OCOc6c5
Synonym 1,3-Benzodioxole, 5,5'-(tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl)bis-, [1S-(1α,3aα,4α,6aα)]-, 1H,3H-Furo[3,4-c]furan, tetrahydro-1,4-bis[3,4-(methylenedioxy)phenyl]-, (1S,3aR,4S,6aR)- (8CI), (1S,3aR,4S,6aR)-5,5'-(Tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl)bis[1,3-benzodioxole], 1H,3H-Furo[3,4-c]furan, 1,3-benzodioxole deriv., (+)-Sesamin, Fagarol, NSC 36403, Sesamin, Sesamin, (+)-, Sesavita, Sezamin, Tetrahydro-1,4-bis[3,4-(methylenedioxy)phenyl]-1H,3H-furo[3,4-c]furan, Zengxiaomin, d-Sesamin, δ-Sesamin
Chemical Name or Material Sesamin

Specifications

Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.