missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seryl tRNA synthetase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80765
This item is not returnable.
View return policy
Description
Seryl tRNA synthetase Polyclonal specifically detects Seryl tRNA synthetase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Seryl tRNA synthetase | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| P49591 | |
| SARS | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NVLSFDDLTADALANLKVSQIKKVRLLIDEAILKCDAERIKLEAERFENLREIGNLLHPSVPISNDEDVDNKVERIWGDCTVRKKYSHVDLVVMVDGFEGEKGAVVAGSRGYFLKGVLVFLEQALIQYAL | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6301 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 6.1.1.11, FLJ36399, serine-tRNA ligase, Serine--tRNA ligase, SERRS, SERSserine tRNA ligase 1, cytoplasmic, Seryl-tRNA Ser/Sec synthetase, seryl-tRNA synthetase, seryl-tRNA synthetase, cytoplasmic, Seryl-tRNA(Ser/Sec) synthetase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of human Seryl tRNA synthetase antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction