missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seryl tRNA synthetase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35334-100ul
This item is not returnable.
View return policy
Description
Seryl tRNA synthetase Polyclonal antibody specifically detects Seryl tRNA synthetase in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Seryl tRNA synthetase | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| EC 6.1.1.11, FLJ36399, serine-tRNA ligase, Serine--tRNA ligase, SERRS, SERSserine tRNA ligase 1, cytoplasmic, Seryl-tRNA Ser/Sec synthetase, seryl-tRNA synthetase, seryl-tRNA synthetase, cytoplasmic, Seryl-tRNA(Ser/Sec) synthetase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human Seryl tRNA synthetase (NP_006504.2).,, Sequence:, MVLDLDLFRVDKGGDPALIRETQEKRFKDPGLVDQLVKADSEWRRCRFRADNLNKLKNLCSKTIGEKMKKKEPVGDDESVPENVLSFDDLTADALANLKVSQIKKVRLLIDEAILKCDAERIKLEAERFENLREIGNLLHPSVPISNDEDVDNKVERIWGDCTVRKKYSHVDLVVMVDGFEGEKGAVVAGSRGYFLKGVLVFLEQALIQYALRTLGSRGYIPIYTPFFMRKEVMQEVAQLSQFDEELYKVIGKGSEKSDDNSYDEKYLIATSEQPIAALHRDEWLRPEDLPIKYAGLSTC | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6301 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction