missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Seryl tRNA synthetase Antibody (1H4), Novus Biologicals™

Product Code. 18346329 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346329 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346329 Supplier Novus Biologicals™ Supplier No. H00006301M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Seryl tRNA synthetase Monoclonal antibody specifically detects Seryl tRNA synthetase in Human,Mouse,Zebrafish samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),ChIP assay (ChIP),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Seryl tRNA synthetase
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), ChIP Assay, KnockDown, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 1H4
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH00716
Gene Alias EC 6.1.1.11, FLJ36399, serine-tRNA ligase, Serine--tRNA ligase, SERRS, SERSserine tRNA ligase 1, cytoplasmic, Seryl-tRNA Ser/Sec synthetase, seryl-tRNA synthetase, seryl-tRNA synthetase, cytoplasmic, Seryl-tRNA(Ser/Sec) synthetase
Host Species Mouse
Immunogen SARS (AAH00716, 1 a.a. ∽ 514 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MVLDLDLFRVDKGGDPALIRETQEKRFKDPGLVDQLVKADSEWRRCRFRADNLNKLKNLCSKTIGEKMKKKEPVGDDESVPENVLSFDDLTADALANLKVSQIKKVRLLIDEAILKCDAERIKLEAERFENLREIGNLLHPSVPISNDEDVDNKVERIWGDCTVRKKYSHVDLVVMVDGFEGEKGAVVAGSRGYFLKGVLVFLEQALIQYALRTLGSRGYIPIYTPFFMRKEVMQEVAQLSQFDEELYKVIGKGSEKSDDNSYDEKYLIATSEQPIAALHRDEWLRPEDLPIKYAGLSTCFRQEVGSHGRDTRGIFRVHQFEKIEQFVYSSPHDNKSWEMFEEMITTAEEFYQSLGIPYHIVNIVSGSLNHAASKKLDLEAWFPGSGAFRELVSCSNCTDYQARRLRIRYGQTKKMMDKVEFVHMLNATMCATTRTICAILENYQTEKGITVPEKLKEFMPPGLQELIPFVKPAPIEQEPSKKQKKQHEGSKKKAAARDVTLENRLQNMEVTDA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6301
Target Species Human, Mouse, Zebrafish
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.