missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Response Factor SRF Mouse anti-Human, Clone: PCRP-SRF-1F7, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00 - £396.00
Specifications
| Antigen | Serum Response Factor SRF |
|---|---|
| Clone | PCRP-SRF-1F7 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18320802
|
Bio-Techne
NBP3-13785-20UG |
20 μg |
£197.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18392107
|
Novus Biologicals
NBP3-13785-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serum Response Factor SRF Monoclonal antibody specifically detects Serum Response Factor SRF in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| Serum Response Factor SRF | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| P11831 | |
| 6722 | |
| IgG2b | |
| Protein A or G purified |
| PCRP-SRF-1F7 | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| MCM1, serum response factor, serum response factor (c-fos serum response element-binding transcriptionfactor) | |
| Recombinant full-length human Serum Response Factor SRF protein (Uniprot: P11831) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title