missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Response Factor SRF Antibody (PCRP-SRF-1F7) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14188JF635
This item is not returnable.
View return policy
Description
Serum Response Factor SRF Monoclonal antibody specifically detects Serum Response Factor SRF in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| Serum Response Factor SRF | |
| Monoclonal | |
| Janelia Fluor 635 | |
| MCM1, serum response factor, serum response factor (c-fos serum response element-binding transcriptionfactor) | |
| Recombinant full-length human Serum Response Factor SRF protein (Uniprot: P11831) | |
| 0.1 mL | |
| Phospho Specific | |
| 6722 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, ELISA, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-SRF-1F7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction